COMH93D67DEE Combi-Blocks, Inc.

Mequitazine

  • MDL number MFCD01706743
  • Cas Number : 29216-28-2
  • Molecular Weight : 322.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8545
Availability details for
Quantity
  • Availability
  • Add to Cart