COMH93D67DFA Combi-Blocks, Inc.

4-Chlorophenyl chlorothionoformate

  • MDL number MFCD00014465
  • Cas Number : 937-64-4
  • Molecular Weight : 207.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name O-(4-chlorophenyl) chloridothiocarbonate
Inchi Code 1S/C7H4Cl2OS/c8-5-1-3-6(4-2-5)10-7(9)11/h1-4H
Inchi Key BQIABQCJXBELMT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8729
Availability details for
Quantity
  • Availability
  • Add to Cart