COMH93D67E0D Combi-Blocks, Inc.

2,4-Dimethylbenzenesulfonic acid sodium salt monohydrate

  • MDL number MFCD15071080
  • Cas Number : 142063-30-7
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sodium 2,4-dimethylbenzenesulfonate hydrate
Inchi Code 1S/C8H10O3S.Na.H2O/c1-6-3-4-8(7(2)5-6)12(9,10)11;;/h3-5H,1-2H3,(H,9,10,11);;1H2/q;+1;/p-1
Inchi Key LAOKFSDVOMOLJF-UHFFFAOYSA-M

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9086
Availability details for
Quantity
  • Availability
  • Add to Cart