COMH93D67E1C Combi-Blocks, Inc.

2,2,2-Trichloroethyl 3,5-dihydroxybenzoate

  • MDL number MFCD02093462
  • Cas Number : 143330-91-0
  • Molecular Weight : 285.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2,2-trichloroethyl 3,5-dihydroxybenzoate
Inchi Code 1S/C9H7Cl3O4/c10-9(11,12)4-16-8(15)5-1-6(13)3-7(14)2-5/h1-3,13-14H,4H2
Inchi Key UJFDPTHAQUCMRW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9340
Availability details for
Quantity
  • Availability
  • Add to Cart