COMH93D67E3D Combi-Blocks, Inc.

10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid

  • MDL number MFCD28976005
  • Cas Number : 1415334-59-6
  • Molecular Weight : 450.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C32H23BO2/c34-33(35)32-29-17-9-7-15-27(29)31(28-16-8-10-18-30(28)32)26-20-24(22-11-3-1-4-12-22)19-25(21-26)23-13-5-2-6-14-23/h1-21,34-35H
Inchi Key NQOSAYPSOKFYBW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9946
Availability details for
Quantity
  • Availability
  • Add to Cart