COMH93D67E75 Combi-Blocks, Inc.

Pacritinib

  • MDL number MFCD22572772
  • Cas Number : 937272-79-2
  • Molecular Weight : 472.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (E)-4(4)-(2-(pyrrolidin-1-yl)ethoxy)-6,11-dioxa-3-aza-2(4,2)-pyrimidina-1,4(1,3)-dibenzenacyclododecaphan-8-ene
Inchi Code 1S/C28H32N4O3/c1-2-13-32(12-1)14-17-35-27-9-8-25-19-24(27)21-34-16-4-3-15-33-20-22-6-5-7-23(18-22)26-10-11-29-28(30-25)31-26/h3-11,18-19H,1-2,12-17,20-21H2,(H,29,30,31)/b4-3+
Inchi Key HWXVIOGONBBTBY-ONEGZZNKSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2626
Availability details for
Quantity
  • Availability
  • Add to Cart