COMH93D67F03 Combi-Blocks, Inc.

2,5-Dimethoxypyridine-3-carboxylic acid

  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6601
Availability details for
Quantity
  • Availability
  • Add to Cart