CO1H324A5BA6 CombiPhos Catalysts Inc

2,3-DI(PYRIDIN-2-YL)PYRIDINE

  • Cas Number : 1235134-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart