CO1H324A5BA8 CombiPhos Catalysts Inc

2,3-DI(PYRIDIN-4-YL)PYRIDINE

  • Cas Number : 1214365-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart