CO1H324A5BA9 CombiPhos Catalysts Inc

3,4-DI(PYRIDIN-2-YL)PYRIDINE

  • Cas Number : 21605-95-8
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart