CO1H324A5BAA CombiPhos Catalysts Inc

3,4-DI(PYRIDIN-3-YL)PYRIDINE

  • Cas Number : 440112-23-2
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart