CO1H324A5BAB CombiPhos Catalysts Inc

3,4-DI(PYRIDIN-4-YL)PYRIDINE

  • Cas Number : 1214336-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart