CO1H324A5BB0 CombiPhos Catalysts Inc

Dihydrogen dichlorobis(di-tert-butylphosphinito-kP)@dipalladate(2-)

  • Cas Number : 391683-95-7
  • Molecular Weight : 518.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart