CO1H324A5BB6 CombiPhos Catalysts Inc

Bis(1,1-dimethylethyl)[2-[(1,1-dimethylethyl)methylphosphino]ethyl]phosphine

  • Cas Number : 705945-72-8
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart