CO1H324A5BBA CombiPhos Catalysts Inc

Bis(1,1-dimethylethyl)(tetrahydro-2-furanyl)phosphine

  • Cas Number : 1621084-37-4
  • Molecular Weight : 216.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart