CO1H324A5BBE CombiPhos Catalysts Inc

2-(2-Furanyl)pyrazine

  • Cas Number : 32736-95-1
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart