CO1H324A5BC4 CombiPhos Catalysts Inc

2,4-Dimethylpyrimidine-5-boronic acid

  • Cas Number : 2170605-42-0
  • Molecular Weight : 151.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart