CO1H324A5BC8 CombiPhos Catalysts Inc

4-Cyclopropylpyrimidine-5-boronic acid

  • Cas Number : 1428882-14-7
  • Molecular Weight : 163.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart