CO1H324A5BCD CombiPhos Catalysts Inc

3-(Cyclobutyl)thiophene-2-boronic acid

  • Cas Number : 2225173-48-6
  • Molecular Weight : 182.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart