CO1H324A5BD0 CombiPhos Catalysts Inc

3-Phenylthiophene-2-boronic acid

  • Cas Number : 2225155-73-5
  • Molecular Weight : 204.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart