CO1H324A5BD1 CombiPhos Catalysts Inc

2-Methyl-1H-imidazole-4-boronic acid

  • Cas Number : 1609265-50-0
  • Molecular Weight : 125.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart