CO1H324A5BD7 CombiPhos Catalysts Inc

5-(sec-Butyl)thiophene-2-boronic acid

  • Cas Number : 2225153-22-8
  • Molecular Weight : 184.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart