CO1H324A5BDA CombiPhos Catalysts Inc

3-(iso-Propyl)furan-2-boronic acid

  • Cas Number : 2225176-02-1
  • Molecular Weight : 153.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart