CO1H324A5BE1 CombiPhos Catalysts Inc

3-(Cyclohexyl)furan-2-boronic acid

  • Cas Number : 2225170-31-8
  • Molecular Weight : 194.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart