CO1H324A5BEC CombiPhos Catalysts Inc

5-(4-Methylimidazol-1-yl)thiophene-2-boronic acid

  • Cas Number : 2225173-65-7
  • Molecular Weight : 208.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart