CO1H324A5BEE CombiPhos Catalysts Inc

5-(1H-Pyrrol-1-yl)thiophene-2-boronic acid

  • Cas Number : 2225169-86-6
  • Molecular Weight : 193.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart