CO1H324A5BF0 CombiPhos Catalysts Inc

5-(Pyrrolidino)thiophene-2-boronic acid

  • Cas Number : 2225172-95-0
  • Molecular Weight : 197.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart