CO1H324A5BF2 CombiPhos Catalysts Inc

5-(Morpholino)thiophene-2-boronic acid

  • Cas Number : 2225179-56-4
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart