CO1H324A5BF9 CombiPhos Catalysts Inc

5-(3-Methylpiperidin-1-yl)furan-2-boronic acid

  • Cas Number : 2225152-11-2
  • Molecular Weight : 209.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart