CO1H324A5C07 CombiPhos Catalysts Inc

4-(4-Methyl-1H-pyrazol-1-yl)furan-2-boronic acid

  • Cas Number : 2225181-26-8
  • Molecular Weight : 191.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart