CO1H324A5C09 CombiPhos Catalysts Inc

4-(Imidazol-1-yl)furan-2-boronic acid

  • Cas Number : 2225173-68-0
  • Molecular Weight : 177.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart