CO1H324A5C12 CombiPhos Catalysts Inc

4-(Morpholino)thiophene-2-boronic acid

  • Cas Number : 2225180-31-2
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart