CO1H324A5C18 CombiPhos Catalysts Inc

4-(3-Methylpiperidin-1-yl)thiophene-2-boronic acid

  • Cas Number : 2225152-10-1
  • Molecular Weight : 225.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart