CO1H324A5C1C CombiPhos Catalysts Inc

4-(Dimethylamino)thiophene-2-boronic acid

  • Cas Number : 2070922-20-0
  • Molecular Weight : 171.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart