CO1H324A5C20 CombiPhos Catalysts Inc

4-(Methylethylamino)thiophene-2-boronic acid

  • Cas Number : 2225153-48-8
  • Molecular Weight : 185.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart