CO1H324A5C26 CombiPhos Catalysts Inc

1,2,5-Trimethylpyrrole-3-boronic acid

  • Cas Number : 2225174-20-7
  • Molecular Weight : 152.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart