CO1H324A5C27 CombiPhos Catalysts Inc

Pyrrole-3-Boronic Acid

  • Cas Number : 763120-55-4
  • Molecular Weight : 110.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart