CO1H324A5C3F CombiPhos Catalysts Inc

2-(4-Methylpiperidin-1-yl)thiazole-4-boronic acid

  • Cas Number : 2096332-87-3
  • Molecular Weight : 226.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart