CO1H324A5C46 CombiPhos Catalysts Inc

2-Iodo-6-methylpyridine-4-boronic acid

  • Cas Number : 2225154-39-0
  • Molecular Weight : 262.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart