CO1H324A5C53 CombiPhos Catalysts Inc

2-Iodo-6-(trifluoromethyl)pyridine-4-boronic acid

  • Cas Number : 2225181-30-4
  • Molecular Weight : 316.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart