CO1H324A5C56 CombiPhos Catalysts Inc

2-Ethyl-6-(trifluoromethyl)pyridine-4-boronic acid

  • Cas Number : 2225170-32-9
  • Molecular Weight : 218.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart