CO1H324A5C58 CombiPhos Catalysts Inc

2-(iso-Propyl)-6-(trifluoromethyl)pyridine-4-boronic acid

  • Cas Number : 2225173-80-6
  • Molecular Weight : 233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart