CO1H324A5C5A CombiPhos Catalysts Inc

2-(Cyclopropyl)-6-(trifluoromethyl)pyridine-4-boronic acid

  • Cas Number : 1446486-08-3
  • Molecular Weight : 230.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart