CO1H324A5C5F CombiPhos Catalysts Inc

2-(Hydroxy)-6-(trifluoromethyl)pyridine-4-boronic acid

  • Cas Number : 2225155-71-3
  • Molecular Weight : 206.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart