CO1H324A5C6A CombiPhos Catalysts Inc

2-Methoxy-6-cyanopyridine-4-boronic acid

  • Cas Number : 2225170-51-2
  • Molecular Weight : 177.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart