CO1H324A5C6D CombiPhos Catalysts Inc

[2-(METHOXYCARBONYL)-6-IODOPYRIDIN-4-YL]BORONIC ACID

  • Cas Number : 2225154-49-2
  • Molecular Weight : 306.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart