CO1H324A5C74 CombiPhos Catalysts Inc

[2-(METHOXYCARBONYL)-6-(TERT-BUTOXY)PYRIDIN-4-YL]BORONIC ACID

  • Cas Number : 2225173-38-4
  • Molecular Weight : 253.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart