CO1H324A5C7C CombiPhos Catalysts Inc

2-Bromo-6-phenylpyridine-4-boronic acid

  • Cas Number : 2225155-85-9
  • Molecular Weight : 277.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart