CO1H324A5C82 CombiPhos Catalysts Inc

2-Bromo-6-(aminomethyl)pyridine-4-boronic acid

  • Cas Number : 2225173-87-3
  • Molecular Weight : 230.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart