CO1H324A5C83 CombiPhos Catalysts Inc

2-Bromo-6-(3-thienyl)pyridine-4-boronic acid

  • Cas Number : 2225179-71-3
  • Molecular Weight : 283.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart